Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ XRCC3 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA521280

Catalog No. PIPA521280


Only null left
Add to Cart

Description

Description

Recommended positive controls: human XRCC3-transfected 293T. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the RecA/Rad51-related protein family that participates in homologous recombination to maintain chromosome stability and repair DNA damage. This gene functionally complements Chinese hamster irs1SF, a repair-deficient mutant that exhibits hypersensitivity to a number of different DNA-damaging agents and is chromosomally unstable. A rare microsatellite polymorphism in this gene is associated with cancer in patients of varying radiosensitivity. Alternatively spliced transcript variants encoding the same protein have been identified.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

XRCC3
Polyclonal
Unconjugated
XRCC3
4432412E01Rik; AI182522; AW537713; CMM6; DNA repair protein XRCC3; DNA repair protein XRCC3-like; LOC102551085; RAD51 like; X-ray repair complementing defective repair in Chinese hamster cells 3; X-ray repair cross complementing 3; X-ray repair cross-complementing protein 3; XRCC 3; XRCC3
Rabbit
Antigen affinity chromatography
RUO
7517
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot, Immunocytochemistry
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
O43542
XRCC3
Recombinant protein encompassing a sequence within the C-terminus region of human XRCC3. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.