Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
XRCC3 Polyclonal Antibody
XRCC3 Polyclonal Antibody
Supplier: Thermo Scientific PA596066
Description
XRCC3 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Specifications
XRCC3 | |
Polyclonal | |
Unconjugated | |
XRCC3 | |
4432412E01Rik; AI182522; AW537713; CMM6; DNA repair protein XRCC3; DNA repair protein XRCC3-like; LOC102551085; RAD51 like; X-ray repair complementing defective repair in Chinese hamster cells 3; X-ray repair cross complementing 3; X-ray repair cross-complementing protein 3; XRCC 3; XRCC3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
7517 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.84 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O43542 | |
XRCC3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 62-346 of human XRCC3 (NP_0010935881). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction