Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XRCC5/Ku80 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501730914 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-0914 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-0914 Supplier Proteintech Group Inc Supplier No. 163891AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Required for activation of ATM following ionizing radiation. May act by regulating dephosphorylation of ATM.

Specifications

Antigen XRCC5/Ku80
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene XRCC5
Gene Accession No. P13010
Gene Alias 86 kDa subunit of Ku antigen, CTC85, CTCBF, DNA repair protein XRCC5, G22P2, KARP 1, KARP1, Ku80, Ku86, KUB2, NFIV, Nuclear factor IV, Thyroid lupus autoantigen, TLAA, XRCC5
Gene Symbols XRCC5
Host Species Rabbit
Immunogen XRCC5/Ku80 Fusion Protein Ag9454
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7520
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.