Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XRCC5 (Ku86) Mouse anti-Human, Unconjugated, Clone: N9C1, MilliporeSigma™

Catalog No. MABE18922MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE18922MI 25 μL
MABE18921MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABE18922MI Supplier MilliporeSigma Supplier No. MABE189225UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

XRCC5 (Ku86) Monoclonal antibody specifically detects XRCC5 (Ku86) in Human samples. It is validated for Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XRCC5 (Ku86)
Applications Western Blot
Classification Monoclonal
Clone N9C1
Conjugate Unconjugated
Gene Accession No. NP_066964
Gene Alias X-ray repair cross-complementing protein 5;86 kDa subunit of Ku antigen;ATP-dependent DNA helicase 2 subunit 2;ATP-dependent DNA helicase II 80 kDa subunit;CTC box-binding factor 85 kDa subunit;CTC85;CTCBF;DNA repair protein XRCC5;Ku80;Ku86;Lupus Ku autoantigen protein p86;Nuclear factor IV;Thyroid-lupus autoantigen;TLAA;X-ray repair complementing defective repair in Chinese hamster cells 5
Gene Symbols XRCC5, G22P2
Host Species Mouse
Immunogen Full-length human recombinant DNA repair protein XRCC5 (Ku86).
Molecular Weight of Antigen ∼80 kDa observed; 82.71 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G Purified
Quantity 25 μL
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Test Specificity Clone N9C1 is a mouse monoclonal antibody that specifically detects DNA repair protein XRCC5.
Target Species Human
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG1 λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.