Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XRCC5/XRCC6 Mouse anti-Human, Clone: KU729 (PE), Abnova™

Catalog No. 89034311 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-034-311 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-034-311 Supplier Abnova Corporation Supplier No. MAB14210
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against native human XRCC5 and XRCC6.

  • High specificity
  • High reactivity

Specifications

Antigen XRCC5/XRCC6
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone KU729
Conjugate PE
Description Mouse monoclonal antibody raised against native human XRCC5 and XRCC6.
Dilution Flow Cytometry (5 uL/test/million cells in 0.1 mL or 5 uL/100 uL of whole blood) Immunofluorescence (1:50-1:100) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% BSA, 0.05% sodium azide).
Gene Accession No. P12956; P13010
Gene Alias CTC75/CTCBF/G22P1/KU70/ML8/TLAA
Gene Symbols XRCC6
Host Species Mouse
Immunogen Nuclear extract of human HL-60 cells.
Purification Method Protein G purification
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2547/7520
Test Specificity This antibody recognizes a conformational epitope of XRCC5/XRCC6 dimer, which is destroyed during Western blotting.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.