Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XRCC6 Mouse anti-Human, Clone:2F7F5, Abnova™

Catalog No. 89333222 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-222 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-222 Supplier Abnova Corporation Supplier No. MAB16607
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human XRCC6.

The p70/p80 autoantigen is a nuclear complex consisting of two subunits with molecular masses of approximately 70 and 80 kDa. The complex functions as a single-stranded DNA-dependent ATP-dependent helicase. The complex may be involved in the repair of nonhomologous DNA ends such as that required for double-strand break repair, transposition, and V(D)J recombination. High levels of autoantibodies to p70 and p80 have been found in some patients with systemic lupus erythematosus. [provided by RefSeq]

Specifications

Antigen XRCC6
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2F7F5
Conjugate Unconjugated
Description X-ray repair complementing defective repair in Chinese hamster cells 6
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunohistochemistry (1:200-1:1000)
Immunocytochemistry (1:200-1:1000)
Flow Cytometry (1:200-1:400)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene XRCC6
Gene Alias CTC75, CTCBF, G22P1, KU70, ML8, TLAA
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acid 6-214 of human XRCC6 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 2547
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.