Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
XRCC6BP1 Antibody (OTI5F3), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274897AF700
Description
XRCC6BP1 Monoclonal specifically detects XRCC6BP1 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
XRCC6BP1 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ATP23 | |
Full length recombinant protein of human XRCC6BP1 (NP_150592) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI5F3 | |
Western Blot, Immunohistochemistry | |
EC 3.4.24, EC 3.4.24.-, Ku70-binding protein 3, KUB3Ku70 binding protein 3, MGC134817, MGC134818, mitochondrial inner membrane protease ATP23 homolog, XRCC6 binding protein 1, XRCC6-binding protein 1 | |
Mouse | |
Affinity Purified | |
Protein Kinase | |
91419.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction