Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ XRCC6BP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557403
Description
Immunogen sequence: CEDCNGNVSG GFDASTSQIV LCQNNIHNQA HMNRVVTHEL IHAFDHCRAH VDWFTNIRHL ACSEVRAANL SGDCSLVNEI Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 86%.
The protein encoded by this gene is amplified in glioblastomas and interacts with the DNA binding subunit of DNA-dependent protein kinase. This kinase is involved in double-strand break repair (DSB), and higher expression of the encoded protein increases the efficiency of DSB. In addition, comparison to orthologous proteins strongly suggests that this protein is a metalloprotease important in the biosynthesis of mitochondrial ATPase. Several transcript variants encoding different isoforms have been found for this gene.
Specifications
XRCC6BP1 | |
Polyclonal | |
Unconjugated | |
ATP23 | |
1110068E08Rik; 2410012H02Rik; atp23; ATP23 metallopeptidase and ATP synthase assembly factor homolog; Ku70 binding protein 3; Ku70-binding protein 3; KUB3; LOW QUALITY PROTEIN: mitochondrial inner membrane protease ATP23 homolog; mitochondrial inner membrane protease ATP23 homolog; XRCC6 binding protein 1; XRCC6-binding protein 1; Xrcc6bp1; zgc:162885 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
91419 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y6H3 | |
ATP23 | |
Recombinant protein corresponding to Human XRCC6BP1. Recombinant protein control fragment (Product #RP-95848). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction