Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ XRCC6BP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565352
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565352 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565352 Supplier Invitrogen™ Supplier No. PA565352
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EQLQQQHVSCQ VFPERLAQGN PQQGFFSSFF TCNQKCQLRL LKTLETNPYV KLLLDAMKHS GCAVNKDRHF Highest antigen sequence identity to the following orthologs - mouse 68%, rat 69%.

The protein encoded by this gene is amplified in glioblastomas and interacts with the DNA binding subunit of DNA-dependent protein kinase. This kinase is involved in double-strand break repair (DSB), and higher expression of the encoded protein increases the efficiency of DSB. In addition, comparison to orthologous proteins strongly suggests that this protein is a metalloprotease important in the biosynthesis of mitochondrial ATPase. Several transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XRCC6BP1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ATP23
Gene Accession No. Q9Y6H3
Gene Alias 1110068E08Rik; 2410012H02Rik; atp23; ATP23 metallopeptidase and ATP synthase assembly factor homolog; Ku70 binding protein 3; Ku70-binding protein 3; KUB3; LOW QUALITY PROTEIN: mitochondrial inner membrane protease ATP23 homolog; mitochondrial inner membrane protease ATP23 homolog; XRCC6 binding protein 1; XRCC6-binding protein 1; Xrcc6bp1; zgc:162885
Gene Symbols ATP23
Host Species Rabbit
Immunogen Recombinant Human XRCC6BP1. Recombinant protein control fragment (Product #RP-106038).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91419
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.