Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Xrn1 Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB500191DL594
Description
Xrn1 Polyclonal antibody specifically detects Xrn1 in Human, Mouse samples. It is validated for Western BlotSpecifications
Xrn1 | |
Polyclonal | |
Western Blot | |
5'-3' exoribonuclease 1, DKFZp434P0721, DKFZp686B22225, DKFZp686F19113, EC 3.1.11.-, SEP1FLJ41903, Strand-exchange protein 1 homolog | |
The immunogen recognized by this antibody maps to a region between residue 1650 and the C-terminus (residue 1706) of human 5'-3' Exoribonuclease 1 using the numbering given in TrEMBL entry Q6MZS8 (GeneID 54464). | |
0.1 mL | |
Cell Cycle and Replication | |
54464 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 594 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction