Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ XRN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100774
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100774 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100774 Supplier Invitrogen™ Supplier No. PA5100774
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total XRN1.

SEC24C is a member of the SEC24 subfamily of the SEC23/SEC24 family, which is involved in vesicle trafficking. The encoded protein has similarity to yeast Sec24p component of COPII. COPII is the coat protein complex responsible for vesicle budding from the ER. The product of this gene may play a role in shaping the vesicle, as well as in cargo selection and concentration.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XRN1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Xrn1
Gene Accession No. P97789, Q8IZH2
Gene Alias 1-Sep; 5'-3' exoribonuclease 1; 5'-3' exoribonuclease 1-like protein; Dhm2; DKFZp434P0721; DKFZp686B22225; DKFZp686F19113; exo; FLJ41903; LOW QUALITY PROTEIN: 5'-3' exoribonuclease 1; mXrn1; protein Dhm2; RGD1309713; SEP1; strand-exchange protein 1 homolog; XRN1
Gene Symbols Xrn1
Host Species Rabbit
Immunogen A synthesized peptide derived from human XRN1(Accession Q8IZH2), corresponding to amino acid residues M1-H45.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24127, 54464
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.