Learn More
XRN1 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300443A
Description
The recommended shelf life for this product is 1 year from date of receipt.
SEP1 (XRN1) localizes to cytoplasmic foci containing a complex of mRNA-degrading enzymes. In addition to mRNA metabolism, yeast Sep1 has been implicated in a variety of nuclear and cytoplasmic functions, including homologous recombination, meiosis, telomere maintece, and microtubule assembly.SEP1 localizes to cytoplasmic foci containing a complex of mRNA-degrading enzymes. In addition to mRNA metabolism, yeast Sep1 has been implicated in a variety of nuclear and cytoplasmic functions, including homologous recombination, meiosis, telomere maintece, and microtubule assembly.[supplied by OMIM].Specifications
| XRN1 | |
| Polyclonal | |
| Unconjugated | |
| XRN1 | |
| 5′-3′ exoribonuclease 1, DKFZp434P0721, DKFZp686B22225, DKFZp686F19113, FLJ41903, SEP1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 24127, 54464 | |
| 4° C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| P97789, Q8IZH2 | |
| XRN1 | |
| Between 1650 and C-term | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.