Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
XRN2 Mouse anti-Human, Clone:9F7G11, Abnova™
Mouse monoclonal antibody raised against recombinant human XRN2.
Supplier: Abnova Corporation MAB17218
Description
This gene shares similarity with the mouse Dhm1 and the yeast dhp1 gene. The yeast gene is involved in homologous recombination and RNA metabolism, such as RNA synthesis and RNA trafficking. Complementation studies show that Dhm1 has a similar function in mouse as dhp1. The function of the human gene has not yet been determined. Transcript variants encoding different isoforms have been noted for this gene; however, their full-length nature is not known. [provided by RefSeq]Specifications
XRN2 | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) Immunocytochemistry Flow Cytometry The optimal working dilution should be determined by the end user. |
|
XRN2 | |
Recombinant protein corresponding to amino acid 398-547 of human XRN2 from E. coli. | |
22803 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
9F7G11 | |
5′-3′ exoribonuclease 2 | |
In PBS (0.05% sodium azide) | |
Mouse | |
100 μg | |
Human | |
Antibody | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction