Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ XRN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565624
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565624 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565624 Supplier Invitrogen™ Supplier No. PA565624
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: THEPNFTIIRE EFKPNKPKPC GLCNQFGHEV KDCEGLPREK KGKHDELADS LPCAEGEFIF LRLNVLREYL ERELTMASLP FTFDVERSID DWVFMCFFV Highest antigen sequence identity to the following orthologs - mouse 98%, rat 98%.

XRN2 gene encodes a 5'-3' exonuclease that promotes transcription termination at cotranscriptional cleavage sites. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XRN2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Xrn2
Gene Accession No. Q9H0D6
Gene Alias 5'-3' exoribonuclease 2; Dhm1; DHM1-like protein; DHP protein; Protein Dhm1; Xrn2; XRN2b
Gene Symbols Xrn2
Host Species Rabbit
Immunogen Recombinant Human XRN2. Recombinant protein control fragment (Product #RP-104906).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22803
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.