Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ XRRA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565227
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565227 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565227 Supplier Invitrogen™ Supplier No. PA565227
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LTSKRYILRF PALETLMLDD NRLSNPSCFA SLAGLRRLKK LSLDENRIIR IPYLQQVQLY DESVDWNGGR GSPHKEPQFM LQSKPRMLED SDEQLDYTVL PM.

May be involved in the response of cells to X-ray radiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XRRA1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene XRRA1
Gene Accession No. Q6P2D8
Gene Alias AI449753; X-ray radiation resistance associated 1; X-ray radiation resistance associated 1 protein; X-ray radiation resistance-associated protein 1; Xrra1
Gene Symbols XRRA1
Host Species Rabbit
Immunogen Recombinant Human XRRA1. Recombinant protein control fragment (Product #RP-105959).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 143570
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.