Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corporation XSelect CSH C18 XP Column

Catalog No. NC1716299 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Change view
Click to view available options
Length (Metric):
30 mm
50 mm
75 mm
100 mm
150 mm
Diameter (Metric):
2.1 mm
3 mm
4.6 mm
Quantity:
1/Ea.
3/Pk.
22 product options available for selection
Product selection table with 22 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric) Quantity
NC1716299 100 mm 3 mm 1/Ea.
50-179-514 30 mm 2.1 mm 1/Ea.
50-179-517 100 mm 2.1 mm 1/Ea.
50-465-931 150 mm 2.1 mm 3/Pk.
50-179-515 50 mm 2.1 mm 1/Ea.
50-179-253 30 mm 2.1 mm 3/Pk.
50-179-254 50 mm 2.1 mm 3/Pk.
50-179-255 75 mm 2.1 mm 3/Pk.
50-179-256 100 mm 2.1 mm 3/Pk.
50-179-257 30 mm 3 mm 3/Pk.
50-179-258 50 mm 3 mm 3/Pk.
50-179-259 75 mm 3 mm 3/Pk.
50-179-260 100 mm 3 mm 3/Pk.
50-179-516 75 mm 2.1 mm 1/Ea.
50-179-518 30 mm 3 mm 1/Ea.
50-179-519 50 mm 3 mm 1/Ea.
50-179-520 75 mm 3 mm 1/Ea.
50-201-538 30 mm 4.6 mm 1/Ea.
50-201-539 50 mm 4.6 mm 1/Ea.
50-201-540 75 mm 4.6 mm 1/Ea.
50-201-541 100 mm 4.6 mm 1/Ea.
50-465-932 150 mm 3 mm 3/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 22 options available.
22 options
Catalog No. NC1716299 Supplier Waters Corporation Supplier No. 186006107
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

XSelect CSH C18 columns are general purpose C18 columns that provide alternative selectivity compared to other C18 sorbents.

  • Charged Surface Hybrid (CSH) technology enables superior peak shape and increased loading capacity for basic compounds.
  • Packed in ultra-low dispersion hardware, 2.5 μm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.

Specifications

Diameter (Metric) 3 mm
Length (Metric) 100 mm
Max. Pressure 18000 psi (1240 Bar)
Particle Shape Spherical
Particle Size 2.5 μm
Pore Size 130 Å
Quantity 1/Ea.
USP Type L1
Endcapped Yes
Product Type eXtended Performance (XP) Column
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.