Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corporation XSelect HSS T3 XP Column

Catalog No. 50465940 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Change view
Click to view available options
Length (Metric):
30 mm
50 mm
75 mm
100 mm
150 mm
Diameter (Metric):
2.1 mm
3 mm
4.6 mm
Quantity:
1/Ea.
3/Pk.
22 product options available for selection
Product selection table with 22 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric) Quantity
50-465-940 150 mm 3 mm 3/Pk.
50-179-544 30 mm 2.1 mm 1/Ea.
50-179-551 100 mm 3 mm 1/Ea.
50-179-547 100 mm 2.1 mm 1/Ea.
50-179-545 50 mm 2.1 mm 1/Ea.
50-179-285 30 mm 2.1 mm 3/Pk.
50-179-286 50 mm 2.1 mm 3/Pk.
50-179-287 75 mm 2.1 mm 3/Pk.
50-179-288 100 mm 2.1 mm 3/Pk.
50-179-289 30 mm 3 mm 3/Pk.
50-179-290 50 mm 3 mm 3/Pk.
50-179-291 75 mm 3 mm 3/Pk.
50-179-292 100 mm 3 mm 3/Pk.
50-179-546 75 mm 2.1 mm 1/Ea.
50-179-548 30 mm 3 mm 1/Ea.
50-179-549 50 mm 3 mm 1/Ea.
50-179-550 75 mm 3 mm 1/Ea.
50-201-561 30 mm 4.6 mm 1/Ea.
50-201-562 50 mm 4.6 mm 1/Ea.
50-201-563 75 mm 4.6 mm 1/Ea.
50-201-564 100 mm 4.6 mm 1/Ea.
50-465-939 150 mm 2.1 mm 3/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 22 options available.
22 options
Catalog No. 50-465-940 Supplier Waters Corporation Supplier No. 176002900
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The XSelect HSS T3 is a low-coverage C18 phase compatible with 100% aqueous mobile phase and should be your first choice when developing separations for polar and non-polar compounds.

  • The XSelect HSS T3 columns exhibit ultra-low MS bleed and are fully scalable and transferable to UPLC separations.
  • Packed in ultra-low dispersion hardware, 2.5 μm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.

Specifications

Diameter (Metric) 3 mm
Length (Metric) 150 mm
Max. Pressure 18000 psi (1240 Bar)
Particle Shape Spherical
Particle Size 2.5 μm
Pore Size 100 Å
Quantity 3/Pk.
USP Type L1
Endcapped Yes
Product Type eXtended Performance (XP) Column
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.