Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
XTP12 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297943H
Description
XTP12 Polyclonal specifically detects XTP12 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
XTP12 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
C6orf62, chromosome 6 open reading frame 62, dJ30M3.2, HBV X-transactivated protein 12, Nbla00237 | |
Produced in rabbits immunized with E. coli-derived Human XTP12 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
HRP | |
PBS | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
81688 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction