Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ XTP3B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556762
Description
Immunogen sequence: PYYPVGMGNG TPCSLKQNRP RSSTVMYICH PESKHEILSV AEVTTCEYEV VILTPLLCSH PKYRFRASPV NDIFCQSLPG S Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
Specifications
| XTP3B | |
| Polyclonal | |
| Unconjugated | |
| ERLEC1 | |
| 4933407N01Rik; C2orf30; cancer invasion and metastasis-related; CIM; CL24936; CL25084; endoplasmic reticulum lectin 1; epididymis luminal protein 117; ER lectin; Erlec1; Erlectin; erlectin 1; HEL117; RGD1306508; UNQ1878/PRO4321; XTP3-B; XTP3TPB; XTP3-transactivated gene B protein; XTP3-transactivated protein B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27248, 289874, 66753 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8VEH8, Q96DZ1 | |
| ERLEC1 | |
| Recombinant protein corresponding to Human XTP3B. Recombinant protein control fragment (Product #RP-95977). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction