Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
XTP3TPA Antibody (03), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306606R
Description
XTP3TPA Monoclonal antibody specifically detects XTP3TPA in Human samples. It is validated for ELISASpecifications
XTP3TPA | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
03 | |
ELISA | |
dCTP pyrophosphatase 1, dCTPase 1, Deoxycytidine-triphosphatase 1, EC 3.6.1.12, MGC5627, RS21C6XTP3-transactivated gene A protein, XTP3TPACDA03, XTP3-transactivated protein A | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human XTP3TPA (NP_077001.1; Met1-Thr170). | |
0.1 mL | |
Proteases & Other Enzymes | |
79077 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction