Learn More
XTP3TPA Antibody, FITC, Novus Biologicals™
SDP
Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals NBP300337F
Catalog No. NB065616
Only null left
Add to Cart
Description
Description
XTP3TPA Polyclonal specifically detects XTP3TPA in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
XTP3TPA
Polyclonal
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
dCTP pyrophosphatase 1, dCTPase 1, Deoxycytidine-triphosphatase 1, EC 3.6.1.12, MGC5627, RS21C6XTP3-transactivated gene A protein, XTP3TPACDA03, XTP3-transactivated protein A
Produced in rabbits immunized with purified, recombinant XTP3TPA (NP_077001.1; Met1-Thr170).
0.1 mL
Proteases & Other Enzymes
79077
Store at 4°C in the dark.
IgG
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
FITC
PBS
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.