Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ XTP3TPA Monoclonal Antibody (03)
GREENER_CHOICE

Catalog No. PIMA529241
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529241 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529241 Supplier Invitrogen™ Supplier No. MA529241
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human XTP3TPA/DCTPP1.

The protein encoded by this gene is dCTP pyrophosphatase, which converts dCTP to dCMP and inorganic pyrophosphate. The encoded protein also displays weak activity against dTTP and dATP, but none against dGTP. This protein may be responsible for eliminating excess dCTP after DNA synthesis and may prevent overmethylation of CpG islands. Three transcript variants, one protein-coding and the other two non-protein coding, have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XTP3TPA
Applications ELISA
Classification Monoclonal
Clone 03
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene DCTPP1
Gene Accession No. Q9H773
Gene Alias 2410015N17Rik; AI854235; CDA03; dCTP pyrophosphatase 1; dCTPase 1; DCTPP1; Deoxycytidine-triphosphatase 1; RS21C6; RS21-C6; Tdrg-TL1; Unknown (protein for MGC:133613); XTP3TPA; XTP3-transactivated gene A protein; XTP3-transactivated gene A protein homolog; XTP3-transactivated protein A
Gene Symbols DCTPP1
Host Species Mouse
Immunogen Recombinant Human XTP3TPA/DCTPP1 Protein (Met1-Thr170).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79077
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.