Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XTP3TPA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542855
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542855 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542855 Supplier Thermo Scientific Supplier No. PA542855
Add to Cart
Add to Cart

XTP3TPA Polyclonal Antibody

XTP3TPA Polyclonal Antibody for Western Blot, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen XTP3TPA
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DCTPP1
Gene Accession No. Q9H773
Gene Alias 2410015N17Rik; AI854235; CDA03; dCTP pyrophosphatase 1; dCTPase 1; DCTPP1; Deoxycytidine-triphosphatase 1; RS21C6; RS21-C6; Tdrg-TL1; Unknown (protein for MGC:133613); XTP3TPA; XTP3-transactivated gene A protein; XTP3-transactivated gene A protein homolog; XTP3-transactivated protein A
Gene Symbols DCTPP1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human XTP3TPA.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79077
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.