Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ XTP3TPA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551750
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551750 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551750 Supplier Invitrogen™ Supplier No. PA551750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RLHAEFAAER DWEQFHQPRN LLLALVGEVG ELAELFQWKT DGEPGPQGWS PRERAALQEE LSDVLIYLVA LAARCRVDLP LAVLSKMDIN RRRYPAHLAR SSSRKYTELP HGAISEDQAV GPADIPCDS Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 78%.

The protein encoded by this gene is dCTP pyrophosphatase, which converts dCTP to dCMP and inorganic pyrophosphate. The encoded protein also displays weak activity against dTTP and dATP, but none against dGTP. This protein may be responsible for eliminating excess dCTP after DNA synthesis and may prevent overmethylation of CpG islands. Three transcript variants, one protein-coding and the other two non-protein coding, have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XTP3TPA
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DCTPP1
Gene Accession No. Q9H773
Gene Alias 2410015N17Rik; AI854235; CDA03; dCTP pyrophosphatase 1; dCTPase 1; DCTPP1; Deoxycytidine-triphosphatase 1; RS21C6; RS21-C6; Tdrg-TL1; Unknown (protein for MGC:133613); XTP3TPA; XTP3-transactivated gene A protein; XTP3-transactivated gene A protein homolog; XTP3-transactivated protein A
Gene Symbols DCTPP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human XTP3TPA. Recombinant protein control fragment (Product #RP-102191).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79077
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.