Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
XTP4 Antibody (OTI2D5), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274904FR
Description
XTP4 Monoclonal specifically detects XTP4 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
XTP4 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
MIEN1 | |
Full length recombinant protein of human XTP4 (NP_115715) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG2b |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI2D5 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
C35, chromosome 17 open reading frame 37, HBV XAg-transactivated protein 4, HBV X-transactivated gene 4 protein, MGC14832, ORB3, protein C17orf37, Protein C35, XTP4RDX12 | |
Mouse | |
Affinity Purified | |
Cancer | |
84299.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction