Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ XYLB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558144
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558144 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558144 Supplier Invitrogen™ Supplier No. PA558144
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DSQHYMALLC FKNGSLMREK IRNESVSRSW SDFSKALQST EMGNGGNLGF YFDVMEITPE IIGRHRFNTE NHKVAAFPGD V Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 81%.

Human xylulokinase-like protein (XYLB), is a 58 kDa protein belongs to a family of enzymes that include fucokinase, gluconokinase, glycerokinase and xylulokinase. XYLB is a key regulator for controlling metabolic disease. XYLB also plays important roles in energy metabolism. XYLB has 2isoformsproduced byalternative splicing with the MW of 58 and 43 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XYLB
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene XYLB
Gene Accession No. O75191, Q3TNA1
Gene Alias AI132223; AI327421; D-xylulokinase; Xylb; Xylulokinase; xylulokinase homolog (H. influenzae); xylulose kinase
Gene Symbols XYLB
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human XYLB. Recombinant protein control fragment (Product #RP-96321).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 102448, 9942
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.