Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylene Cyanol, pure

Catalog No. AC422690500
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
100 g
500 g
Packaging:
Glass bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC422690500 50 g Glass bottle
AC422690050 5 g Glass bottle
AC422690250 25 g Glass bottle
AC422691000 100 g Glass bottle
AC422695000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AC422690500 Supplier Thermo Scientific Chemicals Supplier No. 422690500
Add to Cart

CAS: 4463-44-9 | C25H28N2NaO7S2 | 555.616 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4463-44-9
Molecular Formula C25H28N2NaO7S2
Molecular Weight (g/mol) 555.616
MDL Number MFCD00019481
InChI Key JKPDSWLBCLGXEQ-UHFFFAOYSA-N
Synonym C.I. 43535, Cyanol FF
PubChem CID 131854036
IUPAC Name 4-[[4-(ethylamino)-3-methylphenyl]-(4-ethylimino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]-6-hydroxybenzene-1,3-disulfonic acid;sodium
SMILES CCNC1=C(C=C(C=C1)C(=C2C=CC(=NCC)C(=C2)C)C3=CC(=C(C=C3S(=O)(=O)O)S(=O)(=O)O)O)C.[Na]

Specifications

Dye Content 80% min.
Infrared Spectrum Authentic
Formula Weight 554.61
Grade Extra Pure
Packaging Glass bottle
Color Blue to Red
Quantity 50 g
Physical Form Crystalline Powder
Chemical Name or Material Xylene cyanol, 85%
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 224-728-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.