Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylenecyanol FF, dye content 70%

Catalog No. AAB2153018
Click to view available options
Quantity:
10 g
50 g

CAS: 2650-17-1 | C25H27N2NaO6S2 | 538.61 g/mol

Xylene Cyanol FF is a tracking dye commonly used for nucleic acids. The compound has been used in both electrophoresis polyacrylamide and agarose gels for various evaluations including DNA sequencing. The compound has also been used in a mobility shift analysis of ssDNA. In addition, two highly sensitive spectrophotometric assays have been developed for the detection of platinum (IV) (sc-250738, Platinum(IV) chloride) and Chromium (sc-214714) with the use of Xylene Cyanol FF. As an organic dye-inactivated Sendai virus vaccine, Xylene Cyanol FF displayed strong protection against the virus.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Xylene Cyanol FF is a tracking dye commonly used for nucleic acids. The compound has been used in both electrophoresis polyacrylamide and agarose gels for various evaluations including DNA sequencing. The compound has also been used in a mobility shift analysis of ssDNA. In addition, two highly sensitive spectrophotometric assays have been developed for the detection of platinum (IV) (sc-250738, Platinum(IV) chloride) and Chromium (sc-214714) with the use of Xylene Cyanol FF. As an organic dye-inactivated Sendai virus vaccine, Xylene Cyanol FF displayed strong protection against the virus.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from acids, acid chlorides, acid anhydrides, strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2650-17-1
Molecular Formula C25H27N2NaO6S2
Molecular Weight (g/mol) 538.61
MDL Number MFCD00019481
InChI Key VVLFAAMTGMGYBS-KRQUPCAFSA-M
Synonym Acid blue 147; C.I. 42135
IUPAC Name sodium 4-{[4-(ethylamino)-3-methylphenyl][(1Z,4Z)-4-(ethylimino)-3-methylcyclohexa-2,5-dien-1-ylidene]methyl}-3-sulfobenzene-1-sulfonate
SMILES [Na+].CCNC1=C(C)C=C(C=C1)C(=C1/C=C\C(=N\CC)\C(C)=C1)\C1=C(C=C(C=C1)S([O-])(=O)=O)S(O)(=O)=O

Specifications

Melting Point ∼295°C (decomposition)
Quantity 50 g
Solubility Information Soluble in water.
Formula Weight 538.61
Chemical Name or Material Xylenecyanol FF, dye content 70%
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 220-167-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.