Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylenes, ≥98.5%, ACS Reagent, xylenes plus ethylbenzene basis, Solstice

Catalog No. 60045659 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
20 L
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-659 2.5 L Glass Bottle
60-045-658 1 L Glass Bottle
60-045-660 20 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-045-659 Supplier Solstice Supplier No. 2476422.5L
Add to Cart
Add to Cart

ACS Reagent,≥98.5% xylenes + ethylbenzene basis

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1330-20-7
MDL Number MFCD00077264

Specifications

Melting Point -48°C
Density 0.86g/mL at 25°C
Quantity 2.5 L
Explosion Limit 0.07
Flash Point 25°C (77°F)
Packaging Glass Bottle
Physical Form Liquid
Linear Formula C6H4(CH3)2
Vapor Pressure 18mmHg (37.7°C)
UN Number UN1307
Boiling Point 137°C to 140°C
Formula Weight 106.17g/mol
Percent Purity ≥98.5% xylenes plus ethylbenzene basis
Grade ACS
Chemical Name or Material Xylenes
Show More Show Less
Hazard Statement P210-P260-P280-P301 + P310-P305 + P351 + P338-P370 + P378
Precautionary Statement H226-H304-H312 + H332-H315-H319-H335-H373-H412
DOTInformation Class 3 - PG 3 - Xylenes
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.