Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylenes, 98+%, Extra Dry, mixed isomers, AcroSeal™

Catalog No. p-3759036
Click to view available options
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle

CAS: 1330-20-7 | C8H10

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1330-20-7, 100-41-4
Molecular Formula C8H10
MDL Number MFCD00077264

Specifications

Melting Point -34.0°C
Color Colorless
Density 0.8650g/mL
Boiling Point 136.0°C to 140.0°C
Quantity 100 mL
Flash Point 23°C
Packaging AcroSeal™ Glass Bottle
Physical Form Liquid
Assay Percent Range 98+%
Residue after Evaporation 0.0003% max.
Specific Gravity 0.865
Linear Formula C6H4(CH3)2
Viscosity 0.6 mPa.s (20°C)
Water 0.005% Max. (Coulometric)
Merck Index 15, 10278
Solubility Information Solubility in water: 0.1g/L water (20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol, dmf, methylene chloride, acetone and ether
Formula Weight 106.17
Percent Purity ≥98%
Grade Extra Dry
Chemical Name or Material Xylenes, Mixture of isomers, Extra Dry
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if inhaled.
Causes skin irritation.
Harmful in contact with skin.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.