Learn More
Xylenes, contains ethylbenzene, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Shop All Avantor ProductsDescription
- For Microelectronic Use
Specifications
Specifications
| Quantity | 500 mL |
| Flash Point | 29.00°C |
| Packaging | Narrow Mouth Amber Glass Bottle |
| Specific Gravity | 1L = 0.86kg |
| Molecular Weight (g/mol) | 106.17 |
| Formula Weight | 106.17 |
| Percent Purity | 98.5% (GC) |
| Grade | ACS |
| Chemical Name or Material | Xylenes, Contains ethylbenzene |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.