Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylenes, Histological Grade, Spectrum™ Chemical
SDP

Catalog No. 18614680 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
4 L
20 L
Packaging:
Amber Glass Bottle
Steel Can
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-614-680 4 L Amber Glass Bottle
18-614-679 20 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-614-680 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. HS0074LT
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1330-20-7 | 106.17 g/mol

Chemical Identifiers

CAS 1330-20-7 , 100-41-4
Molecular Weight (g/mol) 106.17

Specifications

CAS Max % 75 to 87%, 13 to 25%
Quantity 4 L
Packaging Amber Glass Bottle
Residue after Evaporation 2e-05
Water 0.0005
Boiling Point 136°C to 141°C
Grade Histological
Chemical Name or Material Xylenes

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.