Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylenes, Reagent, ACS, Spectrum™ Chemical
SDP

Catalog No. 18607765 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Amber Glass Bottle
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-765 100 mL Amber Glass Bottle
18-607-766 1 L Amber Glass Bottle
18-607-772 6 x 500 mL Amber Glass Bottle
18-607-767 4 L Amber Glass Bottle
18-607-768 6 x 1 L Amber Glass Bottle
18-607-769 4 x 4 L Amber Glass Bottle
18-607-770 20 L Amber Glass Bottle
18-607-773 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 missing translation for 'options'
Catalog No. 18-607-765 missing translation for 'mfr' Spectrum Chemical Manufacturing Corporation missing translation for 'supplierNo' X1005100ML
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

X1005, 1330-20-7, C8H10

Spectrum™ Chemical Xylenes, Reagent, ACS is a hydrocarbon that is aromatic and is a mixture of isomers. Used as a solvent that is colorless and has a sweet aroma but is extremely flammable. It is used in the laboratory in conjunction with dry ice to act as a coolant. As an ACS grade Reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 1330-20-7 , 100-41-4
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
InChI Key IVSZLXZYQVIEFR-UHFFFAOYSA-N
IUPAC Name 1,3-xylene
SMILES CC1=CC(C)=CC=C1

Specifications

CAS Max % 75 to 87%, 13 to 25%
Quantity 100 mL
Packaging Amber Glass Bottle
Residue after Evaporation 2e-005
Water 0.0005
Boiling Point 136°C to 141°C
Grade Reagent
Concentration or Composition (by Analyte or Components) 0.985
Chemical Name or Material Xylenes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.