Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Xylenol Orange, 0.2% Alcoholic, Certified, LabChem™
Click to view available options
Quantity:
125 mL
500 mL
Specifications
Specifications
| Chemical Name or Material | Xylenol Orange |
| Name Note | 0.2% Alcoholic |
| CAS Max % | 90.8, 4.6, 4.4, 0.2 |
| CAS | 67-63-0, 67-56-1, 3618-43-7 |
| Linear Formula | C31H28N2Na4O13S |
| Solubility Information | Soluble in water |
| Identification | Passes Test |
| Formula Weight | 760.59 |
| Grade | Certified |
| Packaging | Poly Bottle |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction