Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylobiose 98.0+%, TCI America™
SDP

Catalog No. X00671G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
X00671G 1 g
X0067100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. X00671G Supplier TCI America Supplier No. X00671G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6860-47-5
Molecular Formula C10H18O9
Molecular Weight (g/mol) 282.245
MDL Number MFCD00135984
InChI Key LGQKSQQRKHFMLI-HEMZSNRESA-N
Synonym 4-O-beta-D-Xylopyranosyl-D-xylose
PubChem CID 131674888
IUPAC Name (2S,3S,5R)-2-[(3R,5S)-4,5,6-trihydroxyoxan-3-yl]oxyoxane-3,4,5-triol
SMILES C1C(C(C(C(O1)OC2COC(C(C2O)O)O)O)O)O

Specifications

Color White
Quantity 1 g
Formula Weight 282.25
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Xylobiose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.