Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylose, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18607794 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
5 g
100 g
500 g
1 kg
Packaging:
Amber Glass Bottle

XY110, 58-86-6, C5H10O5

Spectrum™ Chemical Xylose, USP - All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 58-86-6
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
InChI Key SRBFZHDQGSBBOR-UHFFFAOYNA-N
IUPAC Name oxane-2,3,4,5-tetrol
SMILES OC1COC(O)C(O)C1O

Specifications

CAS Max % 1
Ignition Residue 0.0005
Loss on Drying 0.001
Packaging Amber Glass Bottle
Quantity 500 g
Specific Rotation +18.2° to +19.4°
Percent Purity 98 to 102%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.