Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ XYLT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552440
Description
Immunogen sequence: VWTVKILHHW VPVAETKFLV APLTFSNRQP IKPEEALKLH NGPLRNAYME QSFQSLNPVL SLPINPAQVE QARRNAASTG TALEGWLDSL VGGMWTAMDI CATGPTACPV MQTCSQTAWS SFSPDPKSEL G Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.
Catalyzes the first step in biosynthesis of glycosaminoglycan. Transfers D-xylose from UDP-D-xylose to specific serine residues of the core protein. Initial enzyme in the biosynthesis of chondroitin sulfate and dermatan sulfate proteoglycans in fibroblasts and chondrocytes.
Specifications
| XYLT1 | |
| Polyclonal | |
| Unconjugated | |
| Xylt1 | |
| 8030490L12; beta-D-xylosyltransferase 1; DBQD2; Peptide O-xylosyltransferase 1; PXYLT1; Xt1; XTI; XT-I; Xylosyltransferase 1; xylosyltransferase I; xylosyltransferase iota; XYLT1; XYLT-1; XYLTI; xylT-I | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64131 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86Y38 | |
| Xylt1 | |
| Recombinant protein corresponding to Human XYLT1. Recombinant protein control fragment (Product #RP-90075). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction