Learn More
Invitrogen™ XYLT2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA529127
Description
Recommended positive controls: 293T, A431, HeLa, Raji. Predicted reactivity: Mouse (98%), Rat (98%), Zebrafish (84%), Japanese Medaka (82%), Dog (98%), Chicken (93%), Chimpanzee (100%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is an isoform of xylosyltransferase, which belongs to a family of glycosyltransferases. This enzyme transfers xylose from UDP-xylose to specific serine residues of the core protein and initiates the biosynthesis of glycosaminoglycan chains in proteoglycans including chondroitin sulfate, heparan sulfate, heparin and dermatan sulfate. The enzyme activity, which is increased in scleroderma patients, is a diagnostic marker for the determination of sclerotic activity in systemic sclerosis.
Specifications
XYLT2 | |
Polyclonal | |
Unconjugated | |
XYLT2 | |
E030002B02Rik; Peptide O-xylosyltransferase 1; peptide O-xylosyltransferase 2; protein xylosyltransferase; protein xylosyltransferase 2; PXYLT2; SOS; UDP-D-xylose:proteoglycan core protein beta-D-xylosyltransferase; UNQ3058/PRO9878; XT2; XT-II; Xylosyltransferase 2; xylosyltransferase II; Xylt2; xylt-II | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
64132 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q9H1B5 | |
XYLT2 | |
Recombinant fragment corresponding to a region within amino acids 272 and 520 of Human Xylosyltransferase II. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.