Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ XYLT2 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA5121496
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121496 20 μL
PIPA5121495 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121496 Supplier Invitrogen™ Supplier No. PA5121496
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The protein encoded by this gene is an isoform of xylosyltransferase, which belongs to a family of glycosyltransferases. This enzyme transfers xylose from UDP-xylose to specific serine residues of the core protein and initiates the biosynthesis of glycosaminoglycan chains in proteoglycans including chondroitin sulfate, heparan sulfate, heparin and dermatan sulfate. The enzyme activity, which is increased in scleroderma patients, is a diagnostic marker for the determination of sclerotic activity in systemic sclerosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XYLT2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene XYLT2
Gene Accession No. Q9H1B5
Gene Alias E030002B02Rik; Peptide O-xylosyltransferase 1; peptide O-xylosyltransferase 2; protein xylosyltransferase; protein xylosyltransferase 2; PXYLT2; SOS; UDP-D-xylose:proteoglycan core protein beta-D-xylosyltransferase; UNQ3058/PRO9878; XT2; XT-II; Xylosyltransferase 2; xylosyltransferase II; Xylt2; xylt-II
Gene Symbols XYLT2
Host Species Rabbit
Immunogen Phe596-Arg865 (Accession Q9H1B5), with N-terminal His Tag.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64132
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.