Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Xylulokinase/XYLB Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP131095
Description
Xylulokinase/XYLB Polyclonal specifically detects Xylulokinase/XYLB in Human samples. It is validated for Western Blot.Specifications
Xylulokinase/XYLB | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
XYLB | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human XYLB. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Tested in human. Not yet tested in any other species. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
1.0mg/mL | |
Western Blot 1:500-1:3000 | |
EC 2.7.1.17, FLJ10343, FLJ12539, FLJ22075, Xylulokinase, xylulokinase (H. influenzae) homolog, xylulokinase homolog (H. influenzae), xylulose kinase | |
Rabbit | |
58 kDa | |
100 μL | |
Lipid and Metabolism | |
9942 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction