Learn More
Stoelting™ Y Maze
SDP
Stoelting™ Y-maze is similar to the T-maze, but has three identical arms. Gradual turns of the Y-maze decrease learning time as compared to the sharp turns of the T-maze.
$2250.00 - $2600.00
Specifications
For Use With (Application) Neuroscience
View More Specs
Products 2
Catalog Number Mfr. No. Animal Price Quantity & Availability  
Catalog Number Mfr. No. Animal Price Quantity & Availability  
10-000-306
SDP
View Documents
Stoelting™
60180
Mouse
Each for $2,250.00
Only null left
Add to Cart
10-000-329
SDP
View Documents
Stoelting™
60280
Rat
Each for $2,600.00
Only null left
Add to Cart
Description
Description
Specifications
Specifications
Neuroscience
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.