Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ YAE1D1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556688
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556688 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556688 Supplier Invitrogen™ Supplier No. PA556688
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GDVFDEEADE SLLAQREWQS NMQRRVKEGY RDGIDAGKAV TLQQGFNQGY KKGAEVILNY GRLRGTLSAL LSWCH Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 87%.

YAE1D1 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen YAE1D1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene YAE1D1
Gene Accession No. Q9NRH1
Gene Alias C7orf36; GK003; Protein YAE1 homolog; YAE1; Yae1 domain containing 1; yae1 domain-containing protein 1; YAE1D1
Gene Symbols YAE1D1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human YAE1D1. Recombinant protein control fragment (Product #RP-92680).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57002
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.