Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
YAF2 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP204127AF488
Description
YAF2 Polyclonal antibody specifically detects YAF2 in Human samples. It is validated for Western BlotSpecifications
YAF2 | |
Polyclonal | |
Western Blot | |
DKFZp779H1820, MGC41856, YY1 associated factor 2, YY1-associated factor 2 | |
The immunogen recognized by this antibody maps to a region between residue 50 and 100 of human YY1 Associated Factor 2 using the numbering given in entry NP_005739.2 (GeneID 10138). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
10138 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction