Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ YAF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530359
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530359 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530359 Supplier Invitrogen™ Supplier No. PA530359
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, Raji. Predicted reactivity: Chicken (86%), Rhesus Monkey (86%), Bovine (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene interacts with YY1, a zinc finger protein involved in negative regulation of muscle-restricted genes. This gene product itself contains a single N-terminal C2-X10-C2 zinc finger, and in contrast to YY1, is up-regulated during myogenic differentiation. It also facilitates proteolytic cleavage of YY1 by the calcium- activated protease, m-calpain, suggesting a mechanism by which this protein antagonizes the negative effect of YY1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen YAF2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene YAF2
Gene Accession No. Q8IY57
Gene Alias 2810021M11Rik; MGC41856; YAF2; YY1 associated factor 2; YY1-associated factor 2
Gene Symbols YAF2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human YAF2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10138
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.