Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
YAP Rat anti-Mouse, Human, Unconjugated, Clone: 8G5, MilliporeSigma™
Rat Monoclonal Antibody
$208.00 - $492.00
Specifications
| Antigen | YAP |
|---|---|
| Clone | 8G5 |
| Concentration | Please refer to lot specific datasheet. |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
MABS2029-25
|
MilliporeSigma
MABS202925UG |
25 μg |
Each for $208.00
|
|
|||||
|
MABS2029-100
|
MilliporeSigma
MABS2029100UG |
100 μg |
Each for $492.00
|
|
|||||
Description
Anti-YAP, clone 8G5, Cat. No. MABS2029, is a rat monoclonal antibody that detects Transcriptional coactivator YAP1 and has been tested or use in Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, and Western Blotting.Specifications
| YAP | |
| Please refer to lot specific datasheet. | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse, Human | |
| Transcriptional coactivator YAP1;Yes-associated protein 1;Protein yorkie homolog;Yes-associated protein YAP65 homolog | |
| YAP1;YAP65 | |
| IgG2a κ | |
| Protein G purified | |
| Clone 8G5 is a rat monoclonal antibody that detects Transcriptional coactivator YAP1. It targets an epitope with in 12 amino acids from the C-terminal region. | |
| 54.46 kDa calculated. |
| 8G5 | |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rat | |
| Epigenetics & Nuclear Function | |
| Purified rat monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
| NP_001123617.1 | |
| KLH-conjugated linear peptide corresponding to 12 amino acids from the C-terminal region of human Transcriptional coactivator YAP1. | |
| Primary | |
| Stable for 1 year at 2-8°C from date of receipt. | |
| Antibodies |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title