Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
YAP1 Antibody (1A12) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication
$206.50 - $499.50
Specifications
Antigen | YAP1 |
---|---|
Clone | 1A12 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:500-1:2000, Simple Western 20 ug/mL, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-5 ug/mL, Immunoprecipitation reported in scientific literature (PMID 28230103), Immunohistochemistry-Paraffin 1:200-1:1000, Knockout Validated, KnockDown Validated reported in scientific literature (PMID 28230103) |
Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoassay, KnockDown |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP222117SS
![]() |
Novus Biologicals
NBP222117SS |
0.025 mL |
Each for $206.50
|
|
|||||
NBP222117
![]() |
Novus Biologicals
NBP222117 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
YAP1 Monoclonal specifically detects YAP1 in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockout Validated, Knockdown Validated.Specifications
YAP1 | |
1.0 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoassay, KnockDown | |
Unconjugated | |
Mouse | |
Breast Cancer, Cancer, Chromatin Research, Core ESC Like Genes, Signal Transduction, Stem Cell Markers, Transcription Factors and Regulators, Tyrosine Kinases | |
P46937 | |
10413 | |
This YAP1 antibody (1A12) was developed against recombinant human YAP1 protein expressed in E. coli. [UniProt# P46937] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
1A12 | |
Western Blot 1:500-1:2000, Simple Western 20 ug/mL, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-5 ug/mL, Immunoprecipitation reported in scientific literature (PMID 28230103), Immunohistochemistry-Paraffin 1:200-1:1000, Knockout Validated, KnockDown Validated reported in scientific literature (PMID 28230103) | |
Monoclonal | |
Precipitation | |
RUO | |
Human | |
65 kDa Yes-associated protein, COB1, Protein Yorkie Homolog, Transcriptional Coactivator YAP1, YAP, YAP1-2gamma, YAP2, YAP2L, YAP65, YAP65yes-associated protein 2, Yes Associated Protein, Yes-Associated Protein, Yes-associated protein 1, Yes-associated protein 1, 65kDa, yes-associated protein beta, yes-associated protein delta, Yes-Associated Protein YAP65 Homolog, YKI, yorkie homolog | |
YAP1 | |
IgG1 | |
Ammonium sulfate precipitation |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title