Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
YAP1, p Ser127 Antibody (SR2105), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | YAP1 |
---|---|
Clone | SR2105 |
Dilution | Western Blot 1:5000-1:20000, Immunohistochemistry 1:50-1:200 |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Description
YAP1 Monoclonal antibody specifically detects YAP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
YAP1 | |
Western Blot 1:5000-1:20000, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
65 kDa Yes-associated protein, COB1, Protein Yorkie Homolog, Transcriptional Coactivator YAP1, YAP, YAP1-2gamma, YAP2, YAP2L, YAP65, YAP65yes-associated protein 2, Yes Associated Protein, Yes-Associated Protein, Yes-associated protein 1, Yes-associated protein 1, 65kDa, yes-associated protein beta, yes-associated protein delta, Yes-Associated Protein YAP65 Homolog, YKI, yorkie homolog | |
A synthesized peptide derived from human phospho-YAP1 [Ser127] (Uniprot #: P46937) | |
Primary | |
Store at -20 degrees C. Avoid freeze/thaw cycles. |
SR2105 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Breast Cancer, Cancer, Chromatin Research, Core ESC Like Genes, Signal Transduction, Stem Cell Markers, Transcription Factors and Regulators, Tyrosine Kinases | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
10413 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title