Learn More
Invitrogen™ YAP1 (R331W Mutant) Polyclonal Antibody

Description
Centrifuge briefly prior to opening. YAP1 is an oncogene and the YAP1 R331W missense mutation from the original family may be associated with lung adenocarcinoma.
Specifications
Specifications
| Antigen | YAP1 (R331W Mutant) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1.14 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 |
| Gene | YAP1 |
| Gene Accession No. | P46937 |
| Gene Alias | 65 kDa Yes-associated protein; AI325207; cb194; CH211-181P1.5-001; COB1; Protein yorkie homolog; sb:cb194; si:ch211-181p1.5; si:dkey-3b8.3; transcriptional coactivator YAP1; wu:fc18c04; yap; Yap1; YAP2; Yap65; Yes associated protein 1; Yes1 associated transcriptional regulator; yes-associated protein 1; yes-associated protein 2; yes-associated protein YAP65 homolog; yes-associated protein, 65 kDa; Yki; Yorkie; yorkie homolog; zgc:158380; zYAP |
| Gene Symbols | YAP1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.