Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ YARS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553882
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553882 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553882 Supplier Invitrogen™ Supplier No. PA553882
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VLCNLKPQKM RGVESQGMLL CASIEGINRQ VEPLDPPAGS APGEHVFVKG YEKGQPDEEL KPKKKVFEKL QADFKISEEC IAQWKQTNFM TKLGSIS Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 89%.

Aminoacyl-tRNA synthetases catalyze the aminoacylation of tRNA by their cognate amino acid. Because of their central role in linking amino acids with nucleotide triplets contained in tRNAs, aminoacyl-tRNA synthetases are thought to be among the first proteins that appeared in evolution. Tyrosyl-tRNA synthetase belongs to the class I tRNA synthetase family. Cytokine activities have also been observed for the human tyrosyl-tRNA synthetase, after it is split into two parts, an N-terminal fragment that harbors the catalytic site and a C-terminal fragment found only in the mammalian enzyme. The N-terminal fragment is an interleukin-8-like cytokine, whereas the released C-terminal fragment is an EMAP II-like cytokine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen YARS
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene YARS1
Gene Accession No. P54577, Q4KM49, Q91WQ3
Gene Alias AL024047; AU018965; CMTDIC; tyrosine tRNA ligase 1, cytoplasmic; tyrosine--tRNA ligase, cytoplasmic; Tyrosine--tRNA ligase, cytoplasmic, N-terminally processed; tyrosyl trna synthetase; tyrosyl--tRNA ligase; tyrosyl-tRNA synthetase; tyrosyl-tRNA synthetase, cytoplasmic; tyrRS; Yars; YRS; YTS
Gene Symbols YARS1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human YARS. Recombinant protein control fragment (Product #RP-92044).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 107271, 313047, 8565
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.