Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ YBX1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA513307
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA513307 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA513307 Supplier Invitrogen™ Supplier No. PA513307
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with bovine, mouse, rat and Xenopus based on sequence homology.

YBX1 binds to splice sites in pre-mRNA and regulates splice site selection. This protein binds and stabilizes cytoplasmic mRNA and contributes to the regulation of translation by modulating the interaction between the mRNA and eukaryotic initiation factors. It binds to promoters that contain a Y-box (5'-CTGATTGGCCAA-3'), such as HLA class II genes. It regulates the transcription of numerous genes and promotes separation of DNA strands that contain mismatches or are modified by cisplatin. It has endonucleolytic activity and can introduce nicks or breaks into double-stranded DNA (in vitro), and it may play a role in DNA repair.
TRUSTED_SUSTAINABILITY

Specifications

Antigen YBX1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene YBX1
Gene Accession No. P67809
Gene Alias 1700102N10Rik; BP8; BP-8; Byb1; C79409; Cbfa; CBF-A; CCAAT-binding; CCAAT-binding transcription factor 1 subunit A; CCAAT-binding transcription factor I subunit A; CSDA2; CSDB; dbpB; DNA binding protein B; DNA-binding protein B; EF1A; EFI-A; enhancer factor 1 alpha; enhancer factor 1 subunit A; enhancer factor I subunit A; major histocompatibility complex, class II, Y box-binding protein I; MDR NF1; MDR-NF1; MGC104858; MGC110976; MGC117250; Msy1; Msy-1; mYB-1a; Nsep1; NSEP-1; nuclease sensitive element binding protein 1; nuclease-sensitive element-binding protein 1; Y box binding protein 1; Y box protein 1; Y box transcription factor; YB1; YB-1; Y-box binding protein 1; Y-box transcription factor; Y-box-binding protein 1; YBX1
Gene Symbols YBX1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 276-305 amino acids from the C-terminal region of human YBX1.
Purification Method Ammonium Sulfate Precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4904
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.